missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHA2 Monoclonal Antibody (2G12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522265
This item is not returnable.
View return policy
Description
Peptide Sequence: SESLSLVLGK SLDREETAEV NLLLVATDGG KPELTGTVQI LIKVLDVNDN EPTFAQSVYK VKLLENTANG TLVVKLNASD ADEGPNSEIV YSLGSDVSST.
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
Specifications
| PCDHA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y5H9 | |
| PCDHA2 | |
| PCDHA2 (NP_061728, 189 a.a. ~ 288 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2G12 | |
| Unconjugated | |
| PCDHA2 | |
| KIAA0345-like 12; PCDHA2; PCDH-ALPHA2; PCDH-alpha-2; protocadherin alpha 2; protocadherin alpha-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56146 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction