missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19723
This item is not returnable.
View return policy
Description
PCDHA2 Polyclonal specifically detects PCDHA2 in Human samples. It is validated for Western Blot.
Specifications
| PCDHA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| KIAA0345-like 12, PCDH-alpha-2, PCDH-ALPHA2, protocadherin alpha 2, protocadherin alpha-2 | |
| Rabbit | |
| 102 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (88%), Rat (86%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000 | |
| Q9Y5H9 | |
| PCDHA2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human PCDHA2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 56146 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction