missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHA12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583290
This item is not returnable.
View return policy
Description
Immunogen sequence: LNGEISYGIK MILPVSEKCM FSINPDTGEI RIYGELDFEE NNAYEIQVNA IDKGIPSMAG HSMVLVEV Highest antigen sequence indentity to the following orthologs: Mouse - 49%, Rat - 50%.
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
Specifications
| PCDHA12 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA12 | |
| cadherin-related neuronal receptor 5; Cnr5; Crnr5; KIAA0345-like 2; MGC138485; MGC141932; PCDHA12; Pcdha13; PCDH-ALPHA12; PCDH-alpha-12; protocadherin alpha 12; protocadherin alpha 13; protocadherin alpha-12; rCNRv12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UN75 | |
| PCDHA12 | |
| Recombinant protein corresponding to Human PCDHA12. Recombinant protein control fragment (Product #RP-103949). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction