missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94710-0.1ml
This item is not returnable.
View return policy
Description
PCDHA12 Polyclonal antibody specifically detects PCDHA12 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PCDHA12 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| KIAA0345-like 2, MGC138485, MGC141932, PCDH-alpha-12, PCDH-ALPHA12, protocadherin alpha 12, protocadherin alpha-12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 713-792 of human PCDHA12 (NP_114070.1). LTLLLYTALRCSAPPTVSRCAPGKPTLVCSSAVGSWSYSQQRRQRVCSAESPPKTDLMAFSPSLQLSREDCLNPPSEVSY | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 56137 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction