missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548525
This item is not returnable.
View return policy
Description
PCDH9 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the protocadherin family, and cadherin superfamily, of transmembrane proteins containing cadherin domains. These proteins mediate cell adhesion in neural tissues in the presence of calcium. The encoded protein may be involved in signaling at neuronal synaptic junctions. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes multiple cadherin domains and a transmembrane region. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| PCDH9 | |
| Polyclonal | |
| Unconjugated | |
| PCDH9 | |
| cadherin superfamily protein VR4-11; Pcdh9; protocadherin 9; protocadherin-9 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9HC56 | |
| PCDH9 | |
| KLH conjugated synthetic peptide between 1018-1046 amino acids from the C-terminal region of human PCDH9. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction