missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH9 Monoclonal Antibody (7G3F7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531830
This item is not returnable.
View return policy
Description
MA5-31830 has been tested in indirect ELISA.
This gene encodes a member of the protocadherin family, and cadherin superfamily, of transmembrane proteins containing cadherin domains. These proteins mediate cell adhesion in neural tissues in the presence of calcium. The encoded protein may be involved in signaling at neuronal synaptic junctions. Sharing a characteristic with other protocadherin genes, this gene has a notably large exon that encodes multiple cadherin domains and a transmembrane region. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| PCDH9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9HC56 | |
| PCDH9 | |
| Purified recombinant fragment of human PCDH9 (AA: 24-148) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 7G3F7 | |
| Unconjugated | |
| PCDH9 | |
| cadherin superfamily protein VR4-11; Pcdh9; protocadherin 9; protocadherin-9 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction