missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143406
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: zebrafish brain.
Specifications
| PCDH8 | |
| Polyclonal | |
| Unconjugated | |
| Pcdh8 | |
| 1700080P15Rik; Activity-regulated cadherin-like protein; arcadlin; PAPC; Paraxial protocadherin; PCDH8; protocadherin 8; protocadherin-8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| 0 | |
| Pcdh8 | |
| Recombinant protein encompassing a sequence within the center region of zebrafish Pcdh8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction