Learn More
PCDH7 Antibody (OTI1F7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-73269AF700
Product Code. 18159885
  • £621.00 / 0.10mL
Description
PCDH7 Monoclonal specifically detects PCDH7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCDH7
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
PCDH7
Full length recombinant protein of human PCDH7 (NP_115833) produced in HEK293T cells.
0.1 mL
5099.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1F7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
BH-PcdhBHPCDHprotocadherin-7, BH-protocadherin (brain-heart), Brain-heart protocadherin, protocadherin 7
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2b