missing translation for 'onlineSavingsMsg'
Learn More

PCDH20 Antibody (2C3), Novus Biologicals™

Product Code. 18408068 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408068 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408068 Supplier Novus Biologicals Supplier No. H00064881M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PCDH20 Monoclonal antibody specifically detects PCDH20 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDH20
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2C3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_073754
Gene Alias FLJ22218, PCDH13, protocadherin 13, protocadherin 20, protocadherin-13, protocadherin-20
Host Species Mouse
Immunogen PCDH20 (NP_073754, 395 a.a. ∽ 494 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RPPEIVPRYIANEIDGVVYLKELEPVNTPIAFFTIRDPEGKYKVNCYLDGEGPFRLSPYKPYNNEYLLETTKPMDYELQQFYEVAVVAWNSEGFHVKRVI
Purification Method Ascites
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64881
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.