Learn More
PCDH19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555648
Description
Immunogen sequence: CSSLTSSLNY FDYHQQTLPL GCRRSESTFL NVENQNTRNT SANHIYHHSF NSQGPQQPDL IINGVPLPET ENYSFDSNYV NSRAHLIKSS STFKDLEGNS LKDSGHEESD QTDSEHDVQR SLYCDTAVND VLNTSVTSMG SQMPD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
| PCDH19 | |
| Polyclonal | |
| Unconjugated | |
| PCDH19 | |
| PCDH19 | |
| Recombit protein corresponding to Human PCDH19 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAB3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only