missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH15 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547865
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Protocadherin-1, rhProtocadherin-8, and rhProtocadherin-10 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintenance of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.
Specifications
| PCDH15 | |
| Polyclonal | |
| Unconjugated | |
| PCDH15 | |
| Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 65217 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96QU1 | |
| PCDH15 | |
| CHO-derived recombinant human Protocadherin-15 Gln27-Ala1376. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction