missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH12 Monoclonal Antibody (868909)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561886
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Protocadherins comprise the largest group within the cadherin family of calcium-dependent cell-cell adhesion molecules. Protocadherin 12 (PCDH12) was initially identified through PCR screening of mouse heart microvascular endothelial cell RNA; further experiments revealed its mRNA to be strongly expressed in highly vascularized organs such as lung and kidney, in addition to glycogen-rich trophoblasts in the placenta. PCDH12-null mice are viable and fertile, but show reduced placental and embryonic sizes when compared to wild-type mice. Further studies showed significant expression changes in 2, 289 genes, including those involved in tissue morphogenesis, angiogenesis, cell-matrix adhesion and migration, immune response and chromatin remodeling.
Specifications
| PCDH12 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O55134 | |
| PCDH12 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Protocadherin-12, Met1-Ala716. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 868909 | |
| Unconjugated | |
| PCDH12 | |
| Pcdh12; Pcdh14; protocadherin 12; protocadherin 14; protocadherin-12; Protocadherin-12, secreted form; UNQ395/PRO731; Vascular cadherin-2; vascular endothelial cadherin 2; vascular endothelial cadherin-2; VECAD2; VE-cad-2; VE-cadherin 2; VE-cadherin-2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 53601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction