missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH11Y Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538781
This item is not returnable.
View return policy
Description
This gene belongs to the protocadherin family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing seven cadherin repeats, a transmembrane domain, and a cytoplasmic tail that differs from those of the classical cadherins. This gene is located on the Y chromosome in a block of X/Y homology and is very closely related to its paralog on the X chromosome. The protein is thought to play a role in cell-cell recognition during development of the central nervous system. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2013].
Specifications
| PCDH11Y | |
| Polyclonal | |
| Unconjugated | |
| PCDH11Y | |
| PCDH11, PCDH22, PCDHY | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83259 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BZA8 | |
| PCDH11Y | |
| A synthetic peptide derived from the internal region of human PCDH-Y | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction