missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH11X/PCDH11Y Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5103355
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PCDH-X/Y.
This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The encoded protein consists of an extracellular domain containing 7 cadherin repeats, a transmembrane domain and a cytoplasmic tail that differs from those of the classical cadherins. The gene is located in a major X/Y block of homology and its most closely related cadherin superfamily member is located in this homologous region on the X chromosome. The protein is thought to play a fundamental role in cell-cell recognition essential for the segmental development and function of the central nervous system. Transcripts arising from alternative splicing encode isoforms with N- and C-terminal variation.
Specifications
| PCDH11X/PCDH11Y | |
| Polyclonal | |
| Unconjugated | |
| PCDH11X | |
| KIAA1326; PCDH11; PCDH11X; PCDH11Y; PCDH22; PCDH-PC; PCDHX; PCDH-X; PCDHY; PCDH-Y; PPP1R119; protein phosphatase 1, regulatory subunit 119; protocadherin 11 X-linked; protocadherin 11 Y-linked; protocadherin 11X; protocadherin 22; protocadherin on the X chromosome; Protocadherin on the Y chromosome; protocadherin prostate cancer; Protocadherin-11; Protocadherin-11 X-linked; protocadherin-11 Y-linked; Protocadherin-22; Protocadherin-PC; protocadherin-S | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27328, 83259 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BZA7, Q9BZA8 | |
| PCDH11X, PCDH11Y | |
| A synthesized peptide derived from human PCDH11Y(Accession Q9BZA8), corresponding to amino acid residues L1304-L1340. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction