missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13645-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the protocadherin subfamily within the cadherin superfamily. The encoded protein is a membrane protein found at cell-cell boundaries. It is involved in neural cell adhesion, suggesting a possible role in neuronal development. The protein includes an extracellular region, containing 7 cadherin-like domains, a transmembrane region and a C-terminal cytoplasmic region. Cells expressing the protein showed cell aggregation activity. Alternative splicing occurs in this gene.Specifications
| PCDH1 | |
| Polyclonal | |
| Unconjugated | |
| PCDH1 | |
| Cadherin like protein 1, PC42, PCDH1, PCDH42, protocadherin 1, Protocadherin 42 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 307481, 5097, 75599 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q08174 | |
| PCDH1 | |
| PCDH1 Fusion Protein Ag4591 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction