Learn More
PCDH1 (Protocadherin 1) Monoclonal Antibody (CSTEM31), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£133.00 - £274.00
Specifications
Antigen | PCDH1 (Protocadherin 1) |
---|---|
Clone | CSTEM31 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953314
|
Invitrogen
53915741 |
25 Tests |
£133.00
Pack of 25 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963314
|
Invitrogen
53915742 |
100 Tests |
£274.00
Pack of 100 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This monoclonal antibody CSTEM31 reacts with human Protocadherin-1, also known as PCDH1 and cadherin-like protein 1. Applications Reported: This CSTEM31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM31 antibody has been pre-diluted and tested by flow cytometric analysis of human iPSCs. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
PCDH1 (Protocadherin 1) Monoclonal antibody specifically detects PCDH1 (Protocadherin 1) in Human samples. It is validated for Flow Cytometry.Specifications
PCDH1 (Protocadherin 1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
2010005A06Rik; AI585920; cadherin-like 1; cadherin-like protein 1; PC42; PCDH1; PCDH42; protocadherin 1; protocadherin 1 (cadherin-like 1); protocadherin 42; protocadherin-1; Protocadherin-42 | |
PCDH1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
PCDH1 |
CSTEM31 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
Q08174 | |
5097 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |