missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102766
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PCCB.
The protein encoded by this gene is a subunit of the propionyl-CoA carboxylase (PCC) enzyme, which is involved in the catabolism of propionyl-CoA. PCC is a mitochondrial enzyme that probably acts as a dodecamer of six alpha subunits and six beta subunits. This gene encodes the beta subunit of PCC. Defects in this gene are a cause of propionic acidemia type II (PA-2).
Specifications
| PCCB | |
| Polyclonal | |
| Unconjugated | |
| PCCB | |
| 1300012P06Rik; AI314687; PCCase subunit beta; Pccb; Propanoyl-CoA:carbon dioxide ligase subunit beta; propionyl CoA carboxylase, beta polypeptide; propionyl Coenzyme A carboxylase, beta polypeptide; propionyl-coA carboxylase B; Propionyl-CoA carboxylase beta chain precursor; propionyl-CoA carboxylase beta chain, mitochondrial; propionyl-CoA carboxylase beta subunit; propionyl-CoA carboxylase subunit beta; R74805 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24624, 5096, 66904 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05166, P07633, Q99MN9 | |
| PCCB | |
| A synthesized peptide derived from human PCCB(Accession P05166), corresponding to amino acid residues L415-A465. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction