missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A9A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66501-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a subunit of the propionyl-CoA carboxylase (PCC) enzyme, which is involved in the catabolism of propionyl-CoA. PCC is a mitochondrial enzyme that probably acts as a dodecamer of six alpha subunits and six beta subunits. This gene encodes the beta subunit of PCC. Defects in this gene are a cause of propionic acidemia type II (PA-2).Specifications
| PCCB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P05166, P07633, P79384, Q99MN9 | |
| PCCB | |
| PCCB Fusion Protein Ag1608 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1A9A5 | |
| Unconjugated | |
| PCCB | |
| PCCase subunit beta, PCCB | |
| Mouse | |
| Protein G | |
| RUO | |
| 100158147, 24624, 5096, 66904 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction