missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21988-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PCCA.
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.Specifications
| PCCA | |
| Polyclonal | |
| Unconjugated | |
| PCCA | |
| PCCA, PCCase subunit alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110821, 5095 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05165, Q91ZA3 | |
| PCCA | |
| PCCA Fusion Protein Ag17271 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction