missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38609-20ul
This item is not returnable.
View return policy
Description
PCCA Polyclonal antibody specifically detects PCCA in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PCCA | |
| Polyclonal | |
| Western Blot 1:1000 - 1:3000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 6.4.1, EC 6.4.1.3, pccA complementation group, PCCase alpha subunit, PCCase subunit alpha, propanoyl-CoA:carbon dioxide ligase alpha subunit, Propanoyl-CoA:carbon dioxide ligase subunit alpha, propionyl CoA carboxylase, alpha polypeptide, propionyl Coenzyme A carboxylase, alpha polypeptide, propionyl-CoA carboxylase alpha chain, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 449-728 of human PCCA (NP_000273.2).,, Sequence:, ISKLITYGSDRTEALKRMADALDNYVIRGVTHNIALLREVIINSRFVKGDISTKFLSDVYPDGFKGHMLTKSEKNQLLAIASSLFVAFQLRAQHFQENSRMPVIKPDIANWELSVKLHDKVHTVVASNNGSVFSVEVDGSKLNVTSTWNLASPLLSVSVDGTQRTVQCLSREAGGNMSIQFLGTVYKVNILTRLAAELNKFMLEKVTEDTSSVLRSPMPGVVVAVSVKPGDAVAEGQEICVIEAMKMQNSMTAGKTGTVKSVHCQAGDTVGEGDLLVELE | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5095 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction