missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19717
This item is not returnable.
View return policy
Description
PCCA Polyclonal specifically detects PCCA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PCCA | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 6.4.1, EC 6.4.1.3, pccA complementation group, PCCase alpha subunit, PCCase subunit alpha, propanoyl-CoA:carbon dioxide ligase alpha subunit, Propanoyl-CoA:carbon dioxide ligase subunit alpha, propionyl CoA carboxylase, alpha polypeptide, propionyl Coenzyme A carboxylase, alpha polypeptide, propionyl-CoA carboxylase alpha chain, mitochondrial | |
| Rabbit | |
| 80 kDa | |
| 0.1 mL | |
| Primary | |
| Species reactivity based on sequence identity: Mouse (86%), Rat (86%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P05165 | |
| PCCA | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PCCA. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5095 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction