missing translation for 'onlineSavingsMsg'
Learn More

PCC0208009, MedChemExpress

Product Code. 16419716
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419716 50 mg 50mg
16490036 2 mg 2mg
16435897 5 mg 5mg
16403168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16419716 Supplier Medchem Express Supplier No. HY100771

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PCC0208009 is a potent IDO inhibitor with an IC50 value of 4.52 nM in HeLa cell. PCC0208009 alleviates neuropathic pain and comorbidities by regulating synaptic plasticity of anterior cingulate cortex (ACC) and amygdala .

PCC0208009 is a potent IDO inhibitor with an IC50 value of 4.52 nM in HeLa cell. PCC0208009 alleviates neuropathic pain and comorbidities by regulating synaptic plasticity of anterior cingulate cortex (ACC) and amygdala .

Specifications

Chemical Name or Material PCC0208009
Physical Form Solid
CAS 1668565-74-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H35N7O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (200.95 mM; Need ultrasonic)
SMILES CC(C)CN(CC(C)C)C1=CC=C(C2=CC=CC=C2C3=NN=NN3)C=C1NC(NC4=CC=C(C)C=C4)=O
Molecular Weight (g/mol) 497.63
Formula Weight 497.63
Percent Purity 98.16%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.