missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCBP4 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20359 can be used with blocking peptide PEP-0479.
MCG10 is a member of the KH-domain RNA-binding protein family. Proteins of this family also referred to as alpha-CPs, bind to RNA with specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. MCG10 is induced by p53 and DNA damage via two p53-responsive promoter elements. Its overexpression in transfected cells results in the suppression of cell proliferation by inducing apoptosis and cell cycle arrest in G2-M. Decreased expression of one isoform of MCG10 has been observed in many forms of lung cancer, leading to speculation that MCG10 can act as a tumor suppressor. Multiple alternatively spliced isoforms of MCG10 have been identified.
Specifications
Specifications
| Antigen | PCBP4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PCBP4 |
| Gene Accession No. | P57723 |
| Gene Alias | 1200003L19Rik; alpha CP-4; alpha-CP4; AlphaCP-4; alphaCP-4 protein; CBP; LIP4; LYST-interacting protein; MCG10; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4; RNA binding protein MCG10 |
| Gene Symbols | PCBP4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?