missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556413
This item is not returnable.
View return policy
Description
Immunogen sequence: TIQGQYAIPH PDQLTKLHQL AMQQTPFPPL GQTNPAFPGE KLPLHSSEEA QNLMGQSSGL DASPPASTHE LTIPNDLIGC IIGRQGTKIN EIR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 78%.
This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| PCBP3 | |
| Polyclonal | |
| Unconjugated | |
| PCBP3 | |
| alpha-CP3; AlphaCP-3; Pcbp3; PCBP3OT; PCBP3-OT1; PCBP3-overlapping transcript; PCBP3-overlapping transcript 1; poly(rC) binding protein 3; poly(rC)-binding protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54039 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57721 | |
| PCBP3 | |
| Recombinant protein corresponding to Human PCBP3. Recombinant protein control fragment (Product #RP-95176). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction