missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31154
This item is not returnable.
View return policy
Description
PCBP2 Polyclonal specifically detects PCBP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PCBP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| PCBP2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PCBP2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5094 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.25mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| alpha-CP2, Heterogeneous nuclear ribonucleoprotein E2, heterogenous nuclear ribonucleoprotein E2, hnRNP E2, hnRNP-E2, HNRPE2, MGC110998, poly(rC) binding protein 2, poly(rC)-binding protein 2 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (93%). Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction