missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCBP2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33275-20ul
This item is not returnable.
View return policy
Description
PCBP2 Monoclonal antibody specifically detects PCBP2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| PCBP2 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| alpha-CP2, Heterogeneous nuclear ribonucleoprotein E2, heterogenous nuclear ribonucleoprotein E2, hnRNP E2, hnRNP-E2, HNRPE2, MGC110998, poly(rC) binding protein 2, poly(rC)-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-350aa of human PCBP2 (NP_001122383.1),, Sequence:, PDLEGPPLEAYTIQGQYAIPQPDLTKLHQLAMQQSHFPMTHGNTGFSGIESSSPEVKGYWGLDASAQTTSHELTIPNDLIGCIIGRQGAKINEIRQMSGAQIKIANPVEGSTDRQVTITGSAASISLAQYL | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5094 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction