missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PCBP2 Monoclonal antibody specifically detects PCBP2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | PCBP2 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6B6 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH01155 |
| Gene Alias | alpha-CP2, Heterogeneous nuclear ribonucleoprotein E2, heterogenous nuclear ribonucleoprotein E2, hnRNP E2, hnRNP-E2, HNRPE2, MGC110998, poly(rC) binding protein 2, poly(rC)-binding protein 2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?