missing translation for 'onlineSavingsMsg'
Learn More

PCBP1 Rabbit anti-Human, Mouse, Rat, Clone: 7M1K0, Novus Biologicals™

Product Code. 18394733
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394733 20 μg 20µL
18365042 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394733 Supplier Bio-Techne Supplier No. NBP31573420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PCBP1 Monoclonal antibody specifically detects PCBP1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBP1
Applications Western Blot
Classification Monoclonal
Clone 7M1K0
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias alpha-CP1, Heterogeneous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein X, hnRNP E1, hnRNP-E1, hnRNP-X, HNRPE1, HNRPX, nucleic acid binding protein sub 2.3, Nucleic acid-binding protein SUB2.3, poly(rC) binding protein 1, poly(rC)-binding protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PCBP1 (Q15365).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5093
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.