missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCBP1 Antibody (2A10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42654MFV500
This item is not returnable.
View return policy
Description
PCBP1 Monoclonal antibody specifically detects PCBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| PCBP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 2A10 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| alpha-CP1, Heterogeneous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein X, hnRNP E1, hnRNP-E1, hnRNP-X, HNRPE1, HNRPX, nucleic acid binding protein sub 2.3, Nucleic acid-binding protein SUB2.3, poly(rC) binding protein 1, poly(rC)-binding protein 1 | |
| Recombinant human PCBP1 (1-163aa) purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 5093 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction