missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCBD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15702-1-AP-150UL
This item is not returnable.
View return policy
Description
PCBD1 encodes pterin-4 alpha-carbinolamine dehydratase, an enzyme involved in phenylalanine hydroxylation. A deficiency of this enzyme leads to hyperphenylalaninemia. The enzyme regulates the homodimerization of the transcription factor hepatocyte nuclear factor 1 (HNF1).Specifications
| PCBD1 | |
| Polyclonal | |
| Unconjugated | |
| PCBD1 | |
| DCOH, Dimerization cofactor of HNF1, PCBD, PCBD1, PCD, PHS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13180, 29700, 5092 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61457, P61458, P61459 | |
| PCBD1 | |
| PCBD1 Fusion Protein Ag8331 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction