missing translation for 'onlineSavingsMsg'
Learn More

PCB-Mix 41 10 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30324613
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324613 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324613 Supplier Dr. Ehrenstorfer Supplier No. DRELA20034100IO
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 32598-13-3, 32598-14-4, 74472-37-0, 31508-00-6, 65510-44-3, 57465-28-8
Quantity 1 mL
Concentration PCB 77 (3,3',4,4'-Tetrachlorobiphenyl) 10 μg/mL∣PCB No. 105 10 μg/mL∣PCB No. 114 10 μg/mL∣PCB No. 118 10 μg/mL∣PCB No. 123 10 μg/mL∣PCB No. 126 10 μg/mL∣PCB No. 156 10 μg/mL∣PCB No. 157 10 μg/mL∣PCB No. 167 10 μg/mL∣PCB No. 169 10 μg/mL∣PCB No. 189 10 μg/mL∣PCB No. 81 10 μg/mL
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.