missing translation for 'onlineSavingsMsg'
Learn More

PCB 101-13C12, TRC

Product Code. 17985015
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
Unit Size:
0.5mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17985015 0.5 mg 0.5mg
17922695 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17985015 Supplier Toronto Research Chemicals Supplier No. TRCP2166280.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104130-39-4
Molecular Formula 13C12 H5 Cl5
Molecular Weight (g/mol) 338.34
Synonym 1,1'-Biphenyl-1,1',2,2',3,3',4,4',5,5',6,6'-13C12, 2,2',4,5,5'-pentachloro- (9CI), 13C12-2,2',4,5,5'-PeCB, 13C12-2,2',4,5,5'-Pentachlorobiphenyl, 13C12-PCB 101, PCB 101-13C12, 2,2',4,5,5'-Pentachloro-1,1'-biphenyl 13C12, 2,2',4,5,5'-PCB 13C12, 2,2',4,5,5'-Pentachlorobiphenyl 13C12, 2,2',4',5,5'-Pentachlorobiphenyl 13C12, 2,4,5,2',5'-Pentachlorobiphenyl 13C12, 2,5,2',4',5'-Pentachlorobiphenyl 13C12, PCB 101 13C12, K 101 13C12, PCB 101 13C12, 2,2',4,5,5'-PentaCB-13C12, PCB No. 101 13C12, 1,1'-Biphenyl-1,1',2,2',3,3',4,4',5,5',6,6'-13C12, 2,2',4,5,5'-pentachloro- (9CI), 13C12-2,2',4,5,5'-PeCB, 13C12-2,2',4,5,5'-Pentachlorobiphenyl, 13C12-PCB 101, PCB 101-13C12, 2,2',4,5,5'-Pentachloro-1,1'-biphenyl 13C12, 2,2',4,5,5'-PCB 13C12, 2,2',4,5,5'-Pentachlorobiphenyl 13C12, 2,2',4',5,5'-Pentachlorobiphenyl 13C12, 2,4,5,2',5'-Pentachlorobiphenyl 13C12, 2,5,2',4',5'-Pentachlorobiphenyl 13C12, CB 101 13C12, K 101 13C12, PCB 101 13C12, 2,2',4,5,5'-PentaCB-13C12
IUPAC Name 1,2,4-trichloro-5-(2,5-dichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
SMILES Cl[13c]1[13cH][13cH][13c](Cl)[13c]([13cH]1)[13c]2[13cH][13c](Cl)[13c](Cl)[13cH][13c]2Cl

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C12H5Cl5/c13-6-1-2-9(14)7(3-6)8-4-11(16)12(17)5-10(8)15/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1
Formula Weight 335.9236
Chemical Name or Material 2,2',4,5,5'-Pentachlorobiphenyl 13C12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.