missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCAF Recombinant Rabbit Monoclonal Antibody (23GB910), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PCAF |
|---|---|
| Clone | 23GB910 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088962
![]() |
Invitrogen™
MA554026 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089370
|
Invitrogen™
MA554027 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCAF Recombinant Monoclonal antibody specifically detects PCAF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PCAF | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92831 | |
| 8850 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB910 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A930006P13Rik; AI461839; AW536563; CAF; CREBBP-associated factor; histone acetylase PCAF; Histone acetyltransferase KAT2B; histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Kat2b; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Pcaf; Spermidine acetyltransferase KAT2B | |
| A synthetic peptide of human PCAF (154-190AA). | |
| Primary | |
| -20°C | |
| KAT2B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title