missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCAF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575181
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PCAF (histone acetyltransferase KAT2B) functions as a histone acetyltransferase (HAT) to promote transcriptional activation. It has significant histone acetyltransferase activity with core histones (H3 and H4) and nucleosome core particles. PCAF also acetylates non-histone proteins such as ACLY, PLK4, and TBX5. It inhibits cell-cycle progression and counteracts the mitogenic activity of the adenoviral oncoprotein E1A. PCAF also acts as a circadian transcriptional coactivator which enhances the activity of the circadian transcriptional activators NPAS2-ARNTL/BMAL1 and CLOCK-ARNTL/BMAL1 heterodimers. It is involved in heart and limb development by mediating acetylation of TBX5 and acetylation regulating nucleocytoplasmic shutting of TBX5.
Specifications
| PCAF | |
| Polyclonal | |
| Unconjugated | |
| KAT2B | |
| A930006P13Rik; AI461839; AW536563; CAF; CREBBP-associated factor; histone acetylase PCAF; Histone acetyltransferase KAT2B; histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Kat2b; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Pcaf; Spermidine acetyltransferase KAT2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18519, 301164, 8850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| Q92831, Q9JHD1 | |
| KAT2B | |
| Synthetic peptide corresponding to amino acids 786-832 of Human PCAF. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction