missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCAF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31192
This item is not returnable.
View return policy
Description
PCAF Polyclonal specifically detects PCAF in Human, Mouse samples. It is validated for Western Blot.
Specifications
| PCAF | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| KAT2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PCAF. The exact sequence is proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Transcription Factors and Regulators | |
| 8850 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.87mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| CREBBP-associated factor, EC 2.3.1.48, GCN5L, Histone acetylase PCAF, histone acetyltransferase KAT2B, Histone acetyltransferase PCAF, K(lysine) acetyltransferase 2B, Lysine acetyltransferase 2B, P, P/CAFGCN5, P300/CBP-associated factorCAF, PCAF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Zebrafish (88%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction