missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PC-12 (rat adrenal gland pheochromocytoma) Nuclear extract lysate, Non-denatured; Abnova
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Western Blotting, Immunoprecipitation
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Adrenal Gland |
| Description | Nuclear extract cell lysate (non-denatured) |
| Lysis Buffer | Buffer A: 10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT. Buffer C: 20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. Buffer D : 20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. |
| Preparation Method | Nuclear extract was prepared by using a modified protocol of Dignam et al. Cells were Harvested and homogenized in Buffer A, and then centrifugated at 25,000 g for 20 minutes to remove cytoplasm and pellet the nuclei. The pellet was re-suspended in Buffer C, and then the suspensions were centrifuged to collect nuclear extract. The supernatant was dialyzed against Buffer D. The dialysate was then centrifuged, divided into aliquots, and stored at -80°C. The protein concentration was determined by the method of Bradford (Bio-Rad protein assay, microplate standard assay). The lysate was adjusted to 2 mg/ml. |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 50 μg |
| Storage Buffer | In Buffer D. |
| Host Species | Rat |
| Concentration | 2.5 mg/mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction