missing translation for 'onlineSavingsMsg'
Learn More

PBXIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP1-83222-25ul

 View more versions of this product

Product Code. 18498720

  • £342.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PBXIP1 Polyclonal specifically detects PBXIP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PBXIP1
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Hematopoietic PBX-interacting protein, HPIPpre-B-cell leukemia transcription factor interacting protein 1, pre-B-cell leukemia homeobox interacting protein 1, pre-B-cell leukemia transcription factor-interacting protein 1
Rabbit
Affinity Purified
RUO
57326
Human
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
PBXIP1
This antibody was developed against Recombinant Protein corresponding to amino acids:SLPVETLGPASRMDPESERALQAPHSPSKTDGKELAGTMDGEGTLFQTESPQSGSILTEETEVKGTLEGDVCGVEPPGPGDTVVQGDLQETTVVTGLGPDTQDLEGQSPPQSLPSTPKAAWIR
25 μL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only