missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX2 Mouse anti-Human, Clone: PCRP-PBX2-1C4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-13938-100UG
This item is not returnable.
View return policy
Description
PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein Array
Specifications
| PBX2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
| P40425 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5089 | |
| Store at 4°C. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
| PCRP-PBX2-1C4 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
| Recombinant full-length human PBX2 protein (Uniprot: P40425) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction