missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX1 Antibody (PSH0-24), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32720
This item is not returnable.
View return policy
Description
PBX1 Monoclonal antibody specifically detects PBX1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PBX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5087 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot | |
| PSH0-24 | |
| Western Blot 1:1000, Immunohistochemistry 1:200 | |
| DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL | |
| Recombinant protein within human PBX1 aa 1-250/ 430. (Uniprot: P40424) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction