missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PBX1 Monoclonal antibody specifically detects PBX1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | PBX1 |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-PBX1-3C8 |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human PBX1 protein (Uniprot: P40424) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?