missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19713
This item is not returnable.
View return policy
Description
PBX1 Polyclonal specifically detects PBX1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| PBX1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Rat (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.7mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:100-1:500 | |
| P40424 | |
| PBX1 | |
| Recombinant protein encompassing a sequence within the center region of human PBX1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5087 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction