missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ PBT Red Screw Caps with Aperture

Brand:  Lenz Laborglasinstrumente™ 01321014

Code : 50

Additional Details : Weight : 0.68000kg

 View more versions of this product

Product Code. 12333379

  • £13.30 / Pack of 10
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PBT
9.5 mm
Screw thread tubes
PBT
1Pack
Cap
Screw Cap
GL 14
DIN thread (GL) and aperture
180°C
PBT
Red
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers