missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ PBT Red Screw Caps

Product Code. 12353379
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 45
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12353379

Brand: Lenz Laborglasinstrumente™ 01320014

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Red
For Use With (Application) Screw thread tubes
Includes DIN thread (GL)
Material PBT
Max. Temperature (Metric) 180°C
Quantity 10Pack
Screw Material PBT
Product Line Cap
Thread Size (Metric) GL 14
Product Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.