missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution, 0.01M, Chem-Lab

Product Code. 10010023
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10010023 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010023 Supplier Chem-Lab Supplier No. CL05.2636.1000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for laboratory use in TMAR, AFNOR determination in water and titrations. Silver Nitrate Solution, 0.01M, Chem-Lab has a concentration of 1.6985g AgNO3 per 1L H2O. It is used in chloride determination, as a precursor to other silver compounds and has disinfectant properties.

Specifications

CAS 7761-88-8, 7732-18-5
Physical Form Liquid
Quantity 1 L
Molecular Formula AgNO3
MDL Number MFCD00003414
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87g/mol
Concentration or Composition (by Analyte or Components) 1.6985g AgNO3 per 1L H2O
Density 1.00g/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.