missing translation for 'onlineSavingsMsg'
Learn More

PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™

Product Code. 10173433
Change view
Click to view available options
Quantity:
4L
20L
Unit Size:
20 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10173433 4L 4 L
10091403 20L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173433 Supplier Fisher BioReagents Supplier No. BP24384
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X PBS buffer solution
  • Eliminates the hassle of dilution from stock solutions or weighing and mixing individual components
  • 1X solution contains 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 0°C
pH 7.3 to 7.5
Physical Form Liquid
Quantity 4L
Packaging Poly Cube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.