missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
Brand: Fisher Bioreagents BP661-10
Code : X2
Additional Details : Weight : 0.14890kg
Quantity | 10L |
---|---|
Packaging | Poly Bottle |
Description
- Provides a convenient way to make 10L of standard 1X phosphate buffer
- Reconstitute contents of bottle in ultrapure water, such as BP2819, to yield a 1X PBS solution containing 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
- Tested for the absence of DNase, RNase, and protease
- PBS is a standard buffer used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Chemical Identifiers
Phosphate Buffered Saline | |
7447-40-7 | |
Solid |
1X Powder Concentrate | |
DNase-, RNase- and Protease-Free | |
PBS |
Specifications
Phosphate Buffered Saline | |
7447-40-7 | |
DNase-, RNase- and Protease-Free | |
14000 to 17800μmhos/cm | |
PBS | |
Poly Bottle | |
98.8 +/- 1.5 or 496 +/- 4g | |
10L |
1X Powder Concentrate | |
0.136 to 0.144M | |
Solid | |
DNase free | |
Sodium Chloride (81%), Sodium Phosphate Dibasic (≈14%), Potassium Phosphate Monobasic (≈3.0%), and Potassium Chloride (≈2%). | |
Protease free | |
White |