Learn More
PBK Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL555-16110-100UL
Description
The PBK antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PBK. This antibody recognizes human, mouse, rat antigen. The PBK antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
| PBK | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 555 | |
| 100 μL | |
| 55872 | |
| IgG |