missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | PBK |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611416
|
Novus Biologicals
NBP2-99521-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639795
|
Novus Biologicals
NBP2-99521-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBK Polyclonal antibody specifically detects PBK in Human samples. It is validated for ELISASpecifications
| PBK | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 55872 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cancer/testis antigen 84, CT84MAPKK-like protein kinase, EC 2.7.12.2, FLJ14385, Nori-3PDZ-binding kinase, PDZ binding kinase, serine/threonine protein kinase, Spermatogenesis-related protein kinase, SPKT-LAK cell-originated protein kinase, TOPKlymphokine-activated killer T-cell-originated protein kinase | |
| Produced in rabbits immunized with purified, recombinant Human PBK (Accession#: NP_060962.2; Met 1-Val 322) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title