missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBK Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99521C
This item is not returnable.
View return policy
Description
PBK Polyclonal specifically detects PBK in Human samples. It is validated for ELISA.
Specifications
| PBK | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 650 | |
| Cancer/testis antigen 84, CT84MAPKK-like protein kinase, EC 2.7.12.2, FLJ14385, Nori-3PDZ-binding kinase, PDZ binding kinase, serine/threonine protein kinase, Spermatogenesis-related protein kinase, SPKT-LAK cell-originated protein kinase, TOPKlymphokine-activated killer T-cell-originated protein kinase | |
| Produced in rabbits immunized with purified, recombinant PBK (NP_060962.2; Met 1-Val 322). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55872 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction