missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PBK Polyclonal specifically detects PBK in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | PBK |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Cancer/testis antigen 84, CT84MAPKK-like protein kinase, EC 2.7.12.2, FLJ14385, Nori-3PDZ-binding kinase, PDZ binding kinase, serine/threonine protein kinase, Spermatogenesis-related protein kinase, SPKT-LAK cell-originated protein kinase, TOPKlymphokine-activated killer T-cell-originated protein kinase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant PBK (NP_060962.2; Met 1-Val 322). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?